Benzotrifluoride ≥98.0%
Supplier: TCI America
|
Danger
|
CAS Number: 98-08-8
MDL Number: MFCD00000372
Molecular Formula: C7H5F3
Molecular Weight: 146.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 102
Melting point (°C): -29
Flash Point (°C): 12
Specific Gravity (20/20): 1.19
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: (Trifluoromethyl)benzene, alpha,alpha,alpha-Trifluorotoluene
Formula:
C₇H₅F₃ Boiling Pt: 102 °C Melting Pt: –29 °C Density: 1.19 (20 °C) Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000372 CAS Number: 98-08-8 EINECS: 202-635-0 UN: 2338 ADR: 3,II Merck Index: 13,01111 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...