1,3,5-Triethylbenzene ≥97.0%

Supplier: TCI America
Warning

T0426-5ML T0426-25ML
TCT0426-005MLEA 100 USD
TCT0426-005ML TCT0426-025ML
1,3,5-Triethylbenzene ≥97.0%
1,3,5-Triethylbenzene

CAS Number: 102-25-0
MDL Number: MFCD00009261
Molecular Formula: C12H18
Molecular Weight: 162.28
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 218
Flash Point (°C): 76
Specific Gravity (20/20): 0.86

Formula: C₁₂H₁₈
MW: 162.27 g/mol
Boiling Pt: 218 °C
Melting Pt: 3 °C
Density: 0.86 (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00009261
CAS Number: 102-25-0
EINECS: 203-017-3
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5b996b005a3316dea1602ae24e58e558ae8af3b&vid=f28e50742bc553371e3cb59d71f7bd3013cd7e1b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR