2,4,6-Trichlorophenol ≥97.0%
Supplier: TCI America
|
Warning
|
T0390-25G
T0390-500G
TCT0390-025GEA
23.41
USD
TCT0390-025G
TCT0390-500G
2,4,6-Trichlorophenol ≥97.0%
2,4,6-Trichlorophenol
CAS Number: 88-06-2
MDL Number: MFCD00002172
Molecular Formula: C6H3Cl3O
Molecular Weight: 197.44
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 120
Melting point (°C): 67
Flash Point (°C): 99
Formula:
C₆H₃Cl₃O MW: 197.45 g/mol Boiling Pt: 120 °C Melting Pt: 67 °C Density: 1.675 g/cm³ (25 °C) Flash Pt: 99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002172 CAS Number: 88-06-2 EINECS: 201-795-9 UN: 2020 ADR: 6.1,III Merck Index: 12,09773 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...