2,4,5-Trichlorophenol ≥95.0%

Supplier: TCI America
Warning

T0389-500G T0389-25G
TCT0389-500GEA 376.38 USD
TCT0389-500G TCT0389-025G
2,4,5-Trichlorophenol ≥95.0%
2,4,5-Trichlorophenol

CAS Number: 95-95-4
MDL Number: MFCD00002170
Molecular Formula: C6H3Cl3O
Molecular Weight: 197.44
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 115
Melting point (°C): 67
Flash Point (°C): 133

Formula: C₆H₃Cl₃O
MW: 197.45 g/mol
Boiling Pt: 115 °C
Melting Pt: 67 °C
Density: 1.68 g/cm³ (20 °C)
Flash Pt: 133 °C
Storage Temperature: Ambient
MDL Number: MFCD00002170
CAS Number: 95-95-4
EINECS: 202-467-8
UN: 3077
ADR: 9,III
Merck Index: 12,09772
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5fbbbffa95e5e7fab15cde0c920da3b8ffbefe5&vid=87f1d8cee9e1cf179b5992753feb0dd3f6b0ffbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR