1,2,3-Tribromopropane ≥97.0%
Supplier: TCI America
T0355-500G
T0355-25G
TCT0355-500GEA
182
USD
TCT0355-500G
TCT0355-025G
1,2,3-Tribromopropane ≥97.0%
1,2,3-Tribromopropane
CAS Number: 96-11-7
MDL Number: MFCD00017884
Molecular Formula: C3H5Br3
Molecular Weight: 280.79
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 220
Melting point (°C): 17
Flash Point (°C): 93
Specific Gravity (20/20): 2.42
Formula:
BrH₂CBrCHCH₂Br MW: 280.78 g/mol Boiling Pt: 220 °C Melting Pt: 17 °C Density: 2.42 (20 °C) Flash Pt: 93 °C |
MDL Number:
MFCD00017884 CAS Number: 96-11-7 EINECS: 202-478-8 Merck Index: 12,09745 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...