1,1,3,3-Tetramethylbutylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2,4,4-Trimethylpentan-2-amine, 2-Amino-2,4,4-trimethylpentane, tert-Octylamine
T0143-25ML
T0143-500ML
TCT0143-025MLEA
23.42
USD
TCT0143-025ML
TCT0143-500ML
1,1,3,3-Tetramethylbutylamine ≥98.0% (by GC, titration analysis)
1,1,3,3-Tetramethylbutylamine
CAS Number: 107-45-9
MDL Number: MFCD00008053
Molecular Formula: C8H19N
Molecular Weight: 129.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 140
Flash Point (°C): 32
Specific Gravity (20/20): 0.78
Other Product Names / Synonyms: 2-Amino-2,4,4-trimethylpentane, tert-Octylamine
Formula:
C₈H₁₉N MW: 129.25 g/mol Boiling Pt: 140 °C Melting Pt: -8 °C Density: 0.78 (20 °C) Flash Pt: 32 °C |
MDL Number:
MFCD00008053 CAS Number: 107-45-9 EINECS: 203-491-1 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...