1,1,3,3-Tetraethoxypropane ≥95.0%

Supplier: TCI America
Warning

Synonyms: Malonaldehyde Bis(diethyl Acetal)

T0093-500ML T0093-25ML T0093-100ML
TCT0093-500MLEA 182 USD
TCT0093-500ML TCT0093-025ML TCT0093-100ML
1,1,3,3-Tetraethoxypropane ≥95.0%
1,1,3,3-Tetraethoxypropane

CAS Number: 122-31-6
MDL Number: MFCD00009240
Molecular Formula: C11H24O4
Molecular Weight: 220.31
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 220
Flash Point (°C): 42
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: Malonaldehyde Bis(diethyl Acetal)

Formula: C₁₁H₂₄O₄
Boiling Pt: 220 °C
Melting Pt: –90 °C
Density: 0.92 (20 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00009240
CAS Number: 122-31-6
EINECS: 204-533-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b349b80f7915e1ad576575426c191e2d7e3c7cb&vid=0b349b80f7915e1ad576575426c191e2d7e3c7cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR