2-Ethylhexyl salicylate ≥98.0%

Supplier: TCI America

Synonyms: Octyl Salicylate, Salicylic acid 2-Ethylhexyl ester, 2-Ethylhexyl 2-hydroxybenzoate, Salicylic acid Octyl ester

S0387-25ML S0387-500ML
TCS0387-025MLEA 27.94 USD
TCS0387-025ML TCS0387-500ML
2-Ethylhexyl salicylate ≥98.0%
2-Ethylhexyl salicylate

CAS Number: 118-60-5
MDL Number: MFCD00053300
Molecular Formula: C15H22O3
Molecular Weight: 250.34
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 190
Flash Point (°C): 113
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: Salicylic Acid 2-Ethylhexyl Ester, Octyl Salicylate, Salicylic Acid Octyl Ester

Formula: C₁₅H₂₂O₃
MW: 250.34 g/mol
Boiling Pt: 190 °C
Density: 1.02 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00053300
CAS Number: 118-60-5
EINECS: 204-263-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edd979e24e10d3c8b6061f03cbb12fd780805697&vid=edd979e24e10d3c8b6061f03cbb12fd780805697&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR