Rifampicin ≥98.0% (by titrimetric analysis)

Supplier: TCI America

R0079-25G R0079-5G
TCR0079-25GEA 298.23 USD
TCR0079-25G TCR0079-5G
Rifampicin ≥98.0% (by titrimetric analysis)
Rifampicin

CAS Number: 13292-46-1
MDL Number: MFCD00151389
Molecular Formula: C43H58N4O12
Molecular Weight: 822.95
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Yellow Red
Melting point (°C): 188
Storage Temperature: 0-10°C


Product Categories: Bioscience, Antibiotic Ingredients
Other Product Names / Synonyms: Rifampin, Rifamycin AMP

Formula: C₄₃H₅₈N₄O₁₂
Melting Pt: 188 °C
MDL Number: MFCD00151389
CAS Number: 13292-46-1
EINECS: 236-312-0
Merck Index: 12,08382
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=caa43f5d24b888262b5bc9149d8ba00c9b1eb325&vid=caa43f5d24b888262b5bc9149d8ba00c9b1eb325&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR