Resveratrol ≥99.0%

Supplier: TCI America

Synonyms: trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol

R0071-1G R0071-5G
TCR0071-1GEA 64.47 USD
TCR0071-1G TCR0071-5G
Resveratrol ≥99.0%
Resveratrol

CAS Number: 501-36-0
MDL Number: MFCD00133799
Molecular Formula: C14H12O3
Molecular Weight: 228.25
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 267
Storage Temperature: 0-10°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals), Histone Deacetylase Activators
Other Product Names / Synonyms: trans-1,2-(3,4',5-Trihydroxydiphenyl)ethylene, 3,4',5-Trihydroxy-trans-stilbene

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Melting Pt: 267 °C
Density: 1.36 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00133799
CAS Number: 501-36-0
Merck Index: 12,08326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38b5115dabc40dd59d338f41e66aa9ddbc6cbab1&vid=38b5115dabc40dd59d338f41e66aa9ddbc6cbab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR