Potassium(3-nitrophenyl)trifluoroborate ≥98.0%

Supplier: TCI America

Synonyms: (3-Nitrophenyl)trifluoroboric acid potassium salt, Potassium trifluoro(3-nitrophenyl)borate

P1819-1G P1819-5G
TCP1819-1GEA 177 USD
TCP1819-1G TCP1819-5G
Potassium(3-nitrophenyl)trifluoroborate ≥98.0%
Potassium(3-nitrophenyl)trifluoroborate

CAS Number: 192863-40-4
MDL Number: MFCD03701627
Molecular Formula: C6H4BF3KNO2
Molecular Weight: 229.01
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 260

Formula: C₆H₄BF₃KNO₂
MW: 229.01 g/mol
Melting Pt: 260 °C
MDL Number: MFCD03701627
CAS Number: 192863-40-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=700aa462ec97fd290e64b511fb68757372e139a7&vid=700aa462ec97fd290e64b511fb68757372e139a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR