Penicillin G sodium salt ≥98.0%
Supplier: TCI America
Synonyms:
6-(2-Phenylacetamido)penicillanic acid sodium salt, Benzylpenicillin sodium salt, Sodium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CAS Number: 69-57-8
MDL Number: MFCD00069666
Molecular Formula: C16H18N2O4S
Molecular Weight: 356.37
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 227
Specific rotation [a]20/D: 300 deg (C=2, H2O)
Product Categories: Bioscience, Antibiotic Ingredients
Other Product Names / Synonyms: Benzylpenicillin Sodium Salt
Formula:
C₁₆H₁₇N₂NaO₄S MW: 356.38 g/mol Melting Pt: 255 Density: 1.38 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00069666 CAS Number: 69-57-8 EINECS: 200-710-2 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...