5-Phenylvaleric acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-Phenylpentanoic acid, 5-Phenylvaleric Acid

P1284-5G P1284-25G
TCP1284-005GEA 38.7 USD
TCP1284-005G TCP1284-025G
5-Phenylvaleric acid ≥99.0% (by titrimetric analysis)
5-Phenylvaleric acid

CAS Number: 2270-20-4
MDL Number: MFCD00004416
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 164
Melting point (°C): 60


Other Product Names / Synonyms: 5-Phenylpentanoic Acid

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 164 °C
Melting Pt: 60 °C
Density: 1.0487 g/cm³ (25 °C)
MDL Number: MFCD00004416
CAS Number: 2270-20-4
EINECS: 218-872-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f072246b7f9ee8b38d229bd880c59775437b3d7&vid=3f072246b7f9ee8b38d229bd880c59775437b3d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR