Methyl-3-pentenoate ≥90.0%
Supplier: TCI America
Synonyms:
trans-3-Pentenoic acid methyll ester, Methyl trans-3-Pentenoate
P1210-5ML
P1210-25ML
TCP1210-005MLEA
74
USD
TCP1210-005ML
TCP1210-025ML
Methyl-3-pentenoate ≥90.0%
Methyl 3-pentenoate
CAS Number: 20515-19-9
MDL Number: MFCD00191576
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 138
Flash Point (°C): 24
Specific Gravity (20/20): 0.93
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: trans-3-Pentenoic Acid Methyl Ester
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 138 °C Density: 0.93 (20 °C) Flash Pt: 24 °C |
MDL Number:
MFCD00191576 CAS Number: 818-58-6 EINECS: 212-453-3 |
Specification Test Results
Purity/Analysis Method | >90.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...