9,10-Phenanthrenequinone ≥95.0%

Supplier: TCI America

Synonyms: Phenanthrene-9,10-dione

P1136-25G P1136-250G
TCP1136-025GEA 53 USD
TCP1136-025G TCP1136-250G
9,10-Phenanthrenequinone ≥95.0%
9,10-Phenanthrenequinone

CAS Number: 84-11-7
MDL Number: MFCD00001163
Molecular Formula: C14H8O2
Molecular Weight: 208.22
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: Reddish Yellow
Melting point (°C): 210
Flash Point (°C): 100


Other Product Names / Synonyms: 9,10-Phenanthraquinone

Formula: C₁₄H₈O₂
MW: 208.22 g/mol
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 210 °C
Density: 1.4045 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00001163
CAS Number: 84-11-7
EINECS: 201-515-5
UN: 3077
ADR: 9,III
Merck Index: 12,07355
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d211ea5e427756e817fcdfd7a4f8f845dba729b1&vid=d211ea5e427756e817fcdfd7a4f8f845dba729b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR