tert-Butyl propiolate ≥98.0%

Supplier: TCI America

Synonyms: 2-Propynoic acid tert-butyl ester, Propiolic acid tert-butyl ester

P1038-5G P1038-25G
TCP1038-005GEA 254.3 USD
TCP1038-005G TCP1038-25G
tert-Butyl propiolate ≥98.0%
tert-Butyl propiolate

CAS Number: 13831-03-3
MDL Number: MFCD00060100
Molecular Formula: C7H10O2
Molecular Weight: 126.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 53
Flash Point (°C): 28
Specific Gravity (20/20): 0.92


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: Propargylic Acid tert-Butyl Ester, Propiolic Acid tert-Butyl Ester, tert-Butyl Propargylate

Formula: C₇H₁₀O₂
MW: 126.16 g/mol
Boiling Pt: 53 °C
Density: 0.92 (20 °C)
Flash Pt: 28 °C
MDL Number: MFCD00060100
CAS Number: 13831-03-3
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32c292c38d3b71a8c799ed2213c4522adb11a9d0&vid=32c292c38d3b71a8c799ed2213c4522adb11a9d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR