4-Propylbenzoic acid ≥99.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
P0945-25G
P0945-500G
TCP0945-025GEA
75
USD
TCP0945-025G
TCP0945-500G
4-Propylbenzoic acid ≥99.0% (by titrimetric analysis)
4-Propylbenzoic acid
CAS Number: 2438-05-3
MDL Number: MFCD00013996
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Melting point (°C): 142
Formula:
C₁₀H₁₂O₂ MW: 164.2 g/mol Boiling Pt: 298 °C (1013 hPa) Melting Pt: 142 °C Density: ~ 1 g/cm³ (20 °C) |
MDL Number:
MFCD00013996 CAS Number: 2438-05-3 |
Specification Test Results
Purity/Analysis Method | >99.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...