(R)-(-)-2-Pentanol ≥98.0%

Supplier: TCI America
Danger

Synonyms: (R)-(-)-Propyl methyl carbinol, (R)-(-)-2-Hydroxy pentane, (R)-(-)-Methyl propyl carbinol

P0744-1ML P0744-5ML
TCP0744-001MLEA 193 USD
TCP0744-001ML TCP0744-5ML
(R)-(-)-2-Pentanol ≥98.0%
(R)-(-)-2-Pentanol

CAS Number: 31087-44-2
MDL Number: MFCD00065953
Molecular Formula: C5H12O
Molecular Weight: 88.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 119
Flash Point (°C): 34
Specific Gravity (20/20): 0.81
Specific rotation [a]20/D: -13.5 deg (neat)

Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 119 °C
Density: 0.81 (20 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00065953
CAS Number: 31087-44-2
UN: 1105
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5992cac1e7a52acd6ef68b3556715737f7bedf57&vid=5992cac1e7a52acd6ef68b3556715737f7bedf57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR