Prednisolone ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Prednisolonum

P0637-1G P0637-5G P0637-25G
TCP0637-001GEA 46.27 USD
TCP0637-001G TCP0637-005G TCP0637-25G
Prednisolone ≥98.0% (by HPLC)
Prednisolone

CAS Number: 50-24-8
MDL Number: MFCD00003649
Molecular Formula: C21H28O5
Molecular Weight: 360.45
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 235
Specific rotation [a]20/D: 100 deg (C=1, Dioxane)


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Bone Metabolism Research Reagents, Antitumor Ingredients, Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients, Pregnanes (Glucocorticoids),
Other Product Names / Synonyms: 11beta,17alpha,21-Trihydroxypregna-1,4-diene-3,20-dione

Formula: C₂₁H₂₈O₅
MW: 360.45 g/mol
Boiling Pt: 571 °C (1013 hPa)
Melting Pt: 235 °C
Density: 1.212 g/cm³ (20 °C)
MDL Number: MFCD00003649
CAS Number: 50-24-8
EINECS: 200-021-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=def5e482872820c4be850ecfb789b3fcb03155b4&vid=def5e482872820c4be850ecfb789b3fcb03155b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR