N-Phenyl-1-naphthylamine ≥98.0%
Supplier: TCI America
Synonyms:
1-Anilinonaphthalene
P0197-500G
P0197-25G
TCP0197-500GEA
147
USD
TCP0197-500G
TCP0197-025G
N-Phenyl-1-naphthylamine ≥98.0%
N-Phenyl-1-naphthylamine
CAS Number: 90-30-2
MDL Number: MFCD00003878
Molecular Formula: C16H13N
Molecular Weight: 219.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 200
Melting point (°C): 60
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Resin Additives
Other Product Names / Synonyms: 1-Anilinonaphthalene
Formula:
C₁₆H₁₃N Boiling Pt: 200 °C Melting Pt: 60 °C |
MDL Number:
MFCD00003878 CAS Number: 90-30-2 EINECS: 201-983-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...