9,10-Phenanthrenequinone ≥99.0%
Supplier: TCI America
CAS Number: 84-11-7
MDL Number: MFCD00001163
Molecular Formula: C14H8O2
Molecular Weight: 208.22
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 210
Flash Point (°C): 241
Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 9,10-Phenanthraquinone
Formula:
C₁₄H₈O₂ Boiling Pt: 360 °C (1013 hPa) Melting Pt: 210 °C Density: 1.4045 g/cm³ (20 °C) Flash Pt: 241 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001163 CAS Number: 84-11-7 EINECS: 201-515-5 UN: 3077 ADR: 9,III Merck Index: 12,07355 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...