1,4-Pentadiene ≥98.0%
Supplier: TCI America
Synonyms:
Penta-1,4-diene
P0038-10ML
P0038-25ML
TCP0038-010MLEA
168.15
USD
TCP0038-010ML
TCP0038-25ML
1,4-Pentadiene ≥98.0%
1,4-Pentadiene
CAS Number: 591-93-5
MDL Number: MFCD00008665
Molecular Formula: C5H8
Molecular Weight: 68.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 26
Flash Point (°C): -20
Specific Gravity (20/20): 0.66
Storage Temperature: <0°C
Formula:
C₅H₈ MW: 68.12 g/mol Boiling Pt: 26 Melting Pt: -148.3 Density: 0.66 (20 °C) Flash Pt: –20 °C |
MDL Number:
MFCD00008665 CAS Number: 591-93-5 EINECS: 209-736-9 UN: 3295 ADR: 3 I |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...