Oleoyl chloride ≥80.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Oleic acid chloride
O0053-25G
O0053-250G
TCO0053-025GEA
83.33
USD
TCO0053-025G
TCO0053-250G
Oleoyl chloride ≥80.0%
Oleoyl chloride
CAS Number: 112-77-6
MDL Number: MFCD00134332
Molecular Formula: C18H33ClO
Molecular Weight: 300.91
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Boiling point (°C): 200
Flash Point (°C): 171
Specific Gravity (20/20): 0.91
Other Product Names / Synonyms: Oleic Acid Chloride
Formula:
C₁₈H₃₃ClO MW: 300.91 g/mol Boiling Pt: 200 °C Melting Pt: –46 °C Density: 0.91 (20 °C) Flash Pt: 171 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134332 CAS Number: 112-77-6 EINECS: 204-005-0 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >80.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...