2-(2-Nitroethoxy)tetrahydropyran ≥85.0%

Supplier: TCI America

N0602-5G N0602-25G
TCN0602-005GEA 117 USD
TCN0602-005G TCN0602-25G
2-(2-Nitroethoxy)tetrahydropyran ≥85.0%
2-(2-Nitroethoxy)tetrahydropyran

CAS Number: 75233-61-3
MDL Number: MFCD00010858
Molecular Formula: C7H13NO4
Molecular Weight: 175.18
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Flash Point (°C): 48
Specific Gravity (20/20): 1.16
Storage Temperature: <0°C


Other Product Names / Synonyms: Tetrahydro-2-(2-nitroethoxy)-2H-pyran

Formula: C₇H₁₃NO₄
Density: 1.16 (20 °C)
Flash Pt: 48 °C
MDL Number: MFCD00010858
CAS Number: 75233-61-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b14f06cfe9cef3dadcfa40bee2f1c04e7d673fec&vid=b14f06cfe9cef3dadcfa40bee2f1c04e7d673fec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >85.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR