5-Nitroquinoline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
N0344-25G
N0344-250G
TCN0344-025GEA
40
USD
TCN0344-025G
TCN0344-250G
5-Nitroquinoline ≥98.0% (by GC, titration analysis)
5-Nitroquinoline
CAS Number: 607-34-1
MDL Number: MFCD00006790
Molecular Formula: C9H6N2O2
Molecular Weight: 174.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 72
Formula:
C₉H₆N₂O₂ Boiling Pt: 323 °C (1013 hPa) Melting Pt: 72 °C |
MDL Number:
MFCD00006790 CAS Number: 607-34-1 EINECS: 210-134-3 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...