1,8-Nonadiene ≥98.0%

Supplier: TCI America
Danger

N0332-5ML N0332-25ML
TCN0332-005MLEA 49 USD
TCN0332-005ML TCN0332-25ML
1,8-Nonadiene ≥98.0%
1,8-Nonadiene

CAS Number: 4900-30-5
MDL Number: MFCD00008669
Molecular Formula: C9H16
Molecular Weight: 124.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 26
Specific Gravity (20/20): 0.74

Formula: C₉H₁₆
Boiling Pt: 132 °C
Density: 0.74 (20 °C)
Flash Pt: 26 °C
MDL Number: MFCD00008669
CAS Number: 4900-30-5
EINECS: 225-527-5
UN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62a6c68918963ea9da6e763a9975440271b3cf1f&vid=62a6c68918963ea9da6e763a9975440271b3cf1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR