Nonadecanoic acid ≥98.0%
Supplier: TCI America
|
Warning
|
CAS Number: 646-30-0
MDL Number: MFCD00002754
Molecular Formula: C19H38O2
Molecular Weight: 298.51
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 238
Melting point (°C): 68
Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Nonadecylic Acid
Formula:
C₁₉H₃₈O₂ Boiling Pt: 238 °C Melting Pt: 68 °C Density: 0.8468 g/cm³ (70 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002754 CAS Number: 646-30-0 EINECS: 211-472-4 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...