6-Methoxysalicylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Hydroxy-6-methoxybenzoic acid, 6-Hydroxy-o-anisic acid

M1941-5G M1941-1G
TCM1941-5GEA 66 USD
TCM1941-5G TCM1941-1G
6-Methoxysalicylic acid ≥98.0% (by GC, titration analysis)
6-Methoxysalicylic acid

CAS Number: 3147-64-6
MDL Number: MFCD00674090
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 137


Other Product Names / Synonyms: 2-Hydroxy-6-methoxybenzoic Acid, 6-Hydroxy-o-anisic Acid

Formula: C₈H₈O₄
MW: 168.15 g/mol
Melting Pt: 137 °C
MDL Number: MFCD00674090
CAS Number: 3147-64-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fda23d7580834dbaa8dbfd810f90a1285223f40&vid=7fda23d7580834dbaa8dbfd810f90a1285223f40&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR