4-Methoxysalicylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Hydroxy-p-anisic acid, 2-Hydroxy-4-methoxybenzoic acid

M1795-25G M1795-5G
TCM1795-25GEA 157 USD
TCM1795-25G TCM1795-5G
4-Methoxysalicylic acid ≥98.0% (by GC, titration analysis)
4-Methoxysalicylic acid

CAS Number: 2237-36-7
MDL Number: MFCD00002450
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 160


Other Product Names / Synonyms: 2-Hydroxy-4-methoxybenzoic Acid, 2-Hydroxy-p-anisic Acid

Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 327 °C (1013 hPa)
Melting Pt: 160 °C
MDL Number: MFCD00002450
CAS Number: 2237-36-7
EINECS: 218-801-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81495d8198c7d4074d442afe74685da23189aa58&vid=6bf461883c4a8cb1682d56ff11d9d26c70614340&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR