N-Methyl-p-toluidine ≥97.0%

Supplier: TCI America
Danger

Synonyms: N,4-Dimethylaniline, p-(Methylamino)toluene, N-Methyl-4-methylaniline, 4-(Methylamino)toluene, 4-Methyl-N-methylaniline

M1315-25ML M1315-250ML
TCM1315-025MLEA 50 USD
TCM1315-025ML TCM1315-250ML
N-Methyl-p-toluidine ≥97.0%
N-Methyl-p-toluidine

CAS Number: 623-08-5
MDL Number: MFCD00025628
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 212
Flash Point (°C): 83
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: N,4-Dimethylaniline, 4-(Methylamino)toluene

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 212 °C
Density: 0.96 (20 °C)
Flash Pt: 83 °C
MDL Number: MFCD00025628
CAS Number: 623-08-5
EINECS: 210-769-6
UN: 2810
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1535366dd5dcdbefd991bc45ce79def0174afc95&vid=1535366dd5dcdbefd991bc45ce79def0174afc95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR