Ethyl-3-mercaptopropionate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 3-mercaptopropanoate, 3-Mercaptopropionic acid ethyl ester

M1038-25G M1038-500G M1038-100G
TCM1038-025GEA 70.1 USD
TCM1038-025G TCM1038-500G TCM1038-100G
Ethyl-3-mercaptopropionate ≥98.0%
Ethyl-3-mercaptopropionate

CAS Number: 5466-06-8
MDL Number: MFCD00004896
Molecular Formula: C5H10O2S
Molecular Weight: 134.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 72
Specific Gravity (20/20): 1.06


Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents
Other Product Names / Synonyms: 3-Mercaptopropionic Acid Ethyl Ester

Formula: C₅H₁₀O₂S
MW: 134.2 g/mol
Boiling Pt: 75 °C (1013 hPa)
Density: 1.06 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00004896
CAS Number: 5466-06-8
EINECS: 226-771-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96f15fbf33471d38769176d89f1a674d5078c380&vid=96f15fbf33471d38769176d89f1a674d5078c380&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR