5-Methylisoxazole ≥95.0%

Supplier: TCI America

M0813-25ML M0813-5ML
TCM0813-025MLEA 234 USD
TCM0813-025ML TCM0813-5ML
5-Methylisoxazole ≥95.0%
5-Methylisoxazole

CAS Number: 5765-44-6
MDL Number: MFCD00003152
Molecular Formula: C4H5NO
Molecular Weight: 83.09
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Specific Gravity (20/20): 1.02

Formula: C₄H₅NO
Boiling Pt: 122 °C
Density: 1.02 (20 °C)
MDL Number: MFCD00003152
CAS Number: 5765-44-6
EINECS: 227-289-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94c300e912fa373be99e5dc9bb50559f48cdf89f&vid=94c300e912fa373be99e5dc9bb50559f48cdf89f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR