4-Methyl-1-pentanol ≥99.0%

Supplier: TCI America

Synonyms: 4-Methylpentan-1-ol

M0775-25ML M0775-10ML
TCM0775-025MLEA 226 USD
TCM0775-025ML TCM0775-010ML
4-Methyl-1-pentanol ≥99.0%
4-Methyl-1-pentanol

CAS Number: 626-89-1
MDL Number: MFCD00002962
Molecular Formula: C6H14O
Molecular Weight: 102.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 152
Flash Point (°C): 57
Specific Gravity (20/20): 0.81

Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 152 °C
Melting Pt: –70 °C
Density: 0.81 (20 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00002962
CAS Number: 626-89-1
EINECS: 210-969-3
UN: 1987
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dec76bd2644fb6e330ce16bd5dc3d9491c2ed70&vid=1dec76bd2644fb6e330ce16bd5dc3d9491c2ed70&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR