6-Methyl-2-heptanone ≥98.0%

Supplier: TCI America

Synonyms: Methyl 4-Methylpentyl ketone, 6-Methylheptan-2-one

M0700-5ML M0700-25ML
TCM0700-005MLEA 104 USD
TCM0700-005ML TCM0700-25ML
6-Methyl-2-heptanone ≥98.0%
6-Methyl-2-heptanone

CAS Number: 928-68-7
MDL Number: MFCD00026527
Molecular Formula: C8H16O
Molecular Weight: 128.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 171
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: Methyl 4-Methylpentyl Ketone

Formula: C₈H₁₆O
MW: 128.21 g/mol
Boiling Pt: 171 °C
Melting Pt: –59 °C
Density: 0.82 (20 °C)
MDL Number: MFCD00026527
CAS Number: 928-68-7
EINECS: 213-179-7
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb2d473907feb93dc34787fdbef727b37b012195&vid=bb2d473907feb93dc34787fdbef727b37b012195&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR