trans-4-Methoxycinnamic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
CAS Number: 830-09-1
MDL Number: MFCD00004398
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 190
Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals, Bioscience, Phenylpropanoids, Analytical Science, Absolute Configuration Determination (Exciton Chirality CD Method)
Other Product Names / Synonyms: O-Methyl-p-coumaric Acid, O-Methyl-p-cumaric Acid
Formula:
C₁₀H₁₀O₃ Melting Pt: 190 °C |
MDL Number:
MFCD00004398 CAS Number: 943-89-5 EINECS: 212-594-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...