Methylmalonic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-Methylmalonic acid
M0568-100G
M0568-25G
TCM0568-100GEA
187.07
USD
TCM0568-100G
TCM0568-025G
Methylmalonic acid ≥98.0% (by titrimetric analysis)
Methylmalonic acid
CAS Number: 516-05-2
MDL Number: MFCD00002656
Molecular Formula: C4H6O4
Molecular Weight: 118.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 135
Formula:
C₄H₆O₄ MW: 118.09 g/mol Melting Pt: 135 °C |
MDL Number:
MFCD00002656 CAS Number: 516-05-2 EINECS: 208-219-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...