4-Methylphthalic anhydride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M0559-500G M0559-25G
TCM0559-500GEA 392 USD
TCM0559-500G TCM0559-025G
4-Methylphthalic anhydride ≥98.0% (by GC, titration analysis)
4-Methylphthalic anhydride

CAS Number: 19438-61-0
MDL Number: MFCD00041856
Molecular Formula: C9H6O3
Molecular Weight: 162.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 299
Melting point (°C): 94
Flash Point (°C): 170

Formula: C₉H₆O₃
MW: 162.14 g/mol
Boiling Pt: 299 °C
Melting Pt: 94 °C
Flash Pt: 170 °C
MDL Number: MFCD00041856
CAS Number: 19438-61-0
EINECS: 243-073-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a289f1fbe6c101cf3878c5fdb256a8c0c59b7347&vid=a289f1fbe6c101cf3878c5fdb256a8c0c59b7347&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR