2-Methyl-2-pentene ≥95.0%

Supplier: TCI America

Synonyms: 1,1-Dimethyl-1-butene

M0541-5ML M0541-25ML
TCM0541-005MLEA 179 USD
TCM0541-005ML TCM0541-025ML
2-Methyl-2-pentene ≥95.0%
2-Methyl-2-pentene

CAS Number: 625-27-4
MDL Number: MFCD00009382
Molecular Formula: C6H12
Molecular Weight: 84.16
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 67
Flash Point (°C): -23
Specific Gravity (20/20): 0.69


Other Product Names / Synonyms: 1,1-Dimethyl-1-butene

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 67 °C
Melting Pt: -135 °C
Density: 0.69 (20 °C)
Flash Pt: -23 °C
Storage Temperature: Ambient
MDL Number: MFCD00009382
CAS Number: 625-27-4
EINECS: 210-883-6
UN: 3295
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2185563c7a6765ddd5e92682e42800c23cb2dea&vid=67432fa4947234a8ed7df69fa33df284b8fc0e17&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR