N-Methylpropionamide ≥99.0%

Supplier: TCI America

M0488-25ML M0488-5ML
TCM0488-025MLEA 199 USD
TCM0488-025ML TCM0488-5ML
N-Methylpropionamide ≥99.0%
N-Methylpropionamide

CAS Number: 1187-58-2
MDL Number: MFCD00009300
Molecular Formula: C4H9NO
Molecular Weight: 87.12
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 147
Flash Point (°C): 105
Specific Gravity (20/20): 0.93

Formula: C₄H₉NO
Boiling Pt: 147 °C
Density: 0.93 (20 °C)
Flash Pt: 105 °C
MDL Number: MFCD00009300
CAS Number: 1187-58-2
EINECS: 214-699-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8801762e45f5f400cca9e97dd9a56345c36bc37&vid=e8801762e45f5f400cca9e97dd9a56345c36bc37&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR