N-Methyl-M-toluidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
CAS Number: 696-44-6
MDL Number: MFCD00035787
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 207
Flash Point (°C): 100
Specific Gravity (20/20): 0.97
Other Product Names / Synonyms: N,3-Dimethylaniline, 3-(Methylamino)toluene
Formula:
C₈H₁₁N Boiling Pt: 207 °C Density: 0.97 (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00035787 CAS Number: 696-44-6 EINECS: 211-795-0 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...