6-Methyl-2-thiouracil ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Methiocil, 2-Mercapto-6-methylpyrimidin-4(1H)-one, Methylthiouracil, MZU, Basethyrin, 4-Hydroxy-2-mercapto 6-methylpyrimidine, Thiothymin
M0443-500G
M0443-25G
TCM0443-500GEA
309
USD
TCM0443-500G
TCM0443-025G
6-Methyl-2-thiouracil ≥98.0% (by titrimetric analysis)
6-Methyl-2-thiouracil
CAS Number: 56-04-2
MDL Number: MFCD00006040
Molecular Formula: C5H6N2OS
Molecular Weight: 142.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: Methiocil
Formula:
C₅H₆N₂OS MW: 142.18 g/mol Boiling Pt: 351 °C (1013 hPa) Density: 1.519 g/cm³ (20 °C) |
MDL Number:
MFCD00006040 CAS Number: 56-04-2 EINECS: 200-252-3 Merck Index: 12,06210 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...