trans-4-Methyl-2-pentene ≥97.0%

Supplier: TCI America

M0394-5ML M0394-25ML
TCM0394-005MLEA 172 USD
TCM0394-005ML TCM0394-025ML
trans-4-Methyl-2-pentene ≥97.0%
trans-4-Methyl-2-pentene

CAS Number: 674-76-0
MDL Number: MFCD00065138
Molecular Formula: C6H12
Molecular Weight: 84.16
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 58
Flash Point (°C): -27
Specific Gravity (20/20): 0.67

Formula: C₆H₁₂
Boiling Pt: 58 °C
Melting Pt: –140.8 °C
Density: 0.67 (20 °C)
Flash Pt: –27 °C
Storage Temperature: Ambient
MDL Number: MFCD00065138
CAS Number: 674-76-0
EINECS: 211-616-6
UN: 3295
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43e61046075f4d1e4e92c1187bcca1e1841b3e0e&vid=d73731c267fe74b9c0b95b8328e7a5f801032679&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR