3-Methyl-3-pentanol ≥98.0%

Supplier: TCI America

Synonyms: Diethylmethylcarbinol

M0387-25ML M0387-100ML
TCM0387-025MLEA 101.09 USD
TCM0387-025ML TCM0387-100ML
3-Methyl-3-pentanol ≥98.0%
3-Methyl-3-pentanol

CAS Number: 77-74-7
MDL Number: MFCD00004483
Molecular Formula: C6H14O
Molecular Weight: 102.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Flash Point (°C): 24
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: Diethylmethylcarbinol

Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 126 °C
Melting Pt: –53 °C
Density: 0.83 (20 °C)
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00004483
CAS Number: 77-74-7
EINECS: 201-053-4
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f45af6dc4662456d35b01d86cd06e5035f5481cf&vid=f45af6dc4662456d35b01d86cd06e5035f5481cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR