3-Methylpentane ≥99.0%

Supplier: TCI America
Danger

M0383-25ML M0383-500ML
TCM0383-025MLEA 56.69 USD
TCM0383-025ML TCM0383-500ML
3-Methylpentane ≥99.0%
3-Methylpentane

CAS Number: 96-14-0
MDL Number: MFCD00009342
Molecular Formula: C6H14
Molecular Weight: 86.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 63
Melting point (°C): -118
Flash Point (°C): -7
Specific Gravity (20/20): 0.66

Formula: C₆H₁₄
MW: 86.18 g/mol
Boiling Pt: 63 °C
Melting Pt: -118 °C
Density: 0.66 (20 °C)
Flash Pt: -7 °C
Storage Temperature: Ambient
MDL Number: MFCD00009342
CAS Number: 96-14-0
EINECS: 202-481-4
UN: 1208
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d84d4094c1939398d1c0249497caec8f8acc89d&vid=1f7444b8f9e76a39206e100610eb44ac31c7491a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR