5-Methyl-3-heptanone ≥95.0%

Supplier: TCI America
Danger

M0335-500ML M0335-25ML
TCM0335-500MLEA 42 USD
TCM0335-500ML TCM0335-025ML
5-Methyl-3-heptanone ≥95.0%
5-Methyl-3-heptanone

CAS Number: 541-85-5
MDL Number: MFCD00009340
Molecular Formula: C8H16O
Molecular Weight: 128.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 157
Melting point (°C): -57
Flash Point (°C): 59
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: Amyl Ethyl Ketone, Ethyl Amyl Ketone

Formula: C₈H₁₆O
Boiling Pt: 157 °C
Melting Pt: –57 °C
Density: 0.82 (20 °C)
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00009340
CAS Number: 541-85-5
EINECS: 208-793-7
UN: 2271
ADR: 3,III
Merck Index: 12,03810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22e364fdcb88d138ad1c3aea621703f79f9515db&vid=a738e0eb97a2936de033f9e6db869a90f83c103c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR