trans-3-Methyl-2-pentene ≥99.0%

Supplier: TCI America

Synonyms: (E)-3-Methyl-2-pentene

M0312-1ML M0312-5ML
TCM0312-001MLEA 80 USD
TCM0312-001ML TCM0312-005ML
trans-3-Methyl-2-pentene ≥99.0%
trans-3-Methyl-2-pentene

CAS Number: 616-12-6
MDL Number: MFCD00009334
Molecular Formula: C6H12
Molecular Weight: 84.16
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 70
Flash Point (°C): -7
Specific Gravity (20/20): 0.70

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 70 °C
Melting Pt: -138.5 °C
Density: 0.70 (20 °C)
Flash Pt: -7 °C
Storage Temperature: Ambient
MDL Number: MFCD00009334
CAS Number: 616-12-6
EINECS: 210-465-3
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR