2-Methyl-1-pentene ≥98.0%

Supplier: TCI America

M0245-25ML M0245-500ML
TCM0245-025MLEA 141 USD
TCM0245-025ML TCM0245-500ML
2-Methyl-1-pentene ≥98.0%
2-Methyl-1-pentene

CAS Number: 763-29-1
MDL Number: MFCD00009405
Molecular Formula: C6H12
Molecular Weight: 84.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 61
Melting point (°C): -136
Flash Point (°C): -28
Specific Gravity (20/20): 0.68

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 61 °C
Melting Pt: –136 °C
Density: 0.68 (20 °C)
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00009405
CAS Number: 763-29-1
EINECS: 212-108-7
UN: 2288
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46d04901ea2c15711fe7830416987d9c4e6a4977&vid=985255819d04ed3f35b63022b25ca08452fd8ee1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR