2-Methoxyethyl acetate ≥98.0%

Supplier: TCI America
Danger

M0113-500ML M0113-25ML
TCM0113-500MLEA 44.75 USD
TCM0113-500ML TCM0113-025ML
2-Methoxyethyl acetate ≥98.0%
2-Methoxyethyl acetate

CAS Number: 110-49-6
MDL Number: MFCD00008719
Molecular Formula: C5H10O3
Molecular Weight: 118.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 143
Melting point (°C): -65
Flash Point (°C): 51
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: Ethylene Glycol Monomethyl Ether Acetate, Acetic Acid 2-Methoxyethyl Ester, Methyl Cellosolve Acetate, Methyl Glycol Acetate

Formula: C₅H₁₀O₃
Boiling Pt: 143 °C
Melting Pt: -65 °C
Density: 1.01 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00008719
CAS Number: 110-49-6
EINECS: 203-772-9
Merck Index: 12,06119
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a79da10fc207e1d48addb5bcd2faa1e327d3cf8e&vid=a79da10fc207e1d48addb5bcd2faa1e327d3cf8e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR