N-Lauroylsarcosine ≥90.0%

Supplier: TCI America

Synonyms: N-Lauroyl-Sar-OH, 2-(N-Methyldodecanamido)acetic acid, N-Dodecanoylsarcosine

L0151-25G L0151-500G L0151-100G
TCL0151-025GEA 22.39 USD
TCL0151-025G TCL0151-500G TCL0151-100G
N-Lauroylsarcosine ≥90.0%
N-Lauroylsarcosine

CAS Number: 97-78-9
MDL Number: MFCD00021749
Molecular Formula: C15H29NO3
Molecular Weight: 271.40
Purity/Analysis Method: >90.0% (GC)
Melting point (°C): 46
Flash Point (°C): 210


Other Product Names / Synonyms: N-Dodecanoylsarcosine

Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COOH
MW: 271.4 g/mol
Melting Pt: 46 °C
Flash Pt: 210 °C
MDL Number: MFCD00021749
CAS Number: 97-78-9
EINECS: 202-608-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7b428c500bc10787606e0dedb3d1759e91cea90&vid=b7b428c500bc10787606e0dedb3d1759e91cea90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR