2,5-Lutidine ≥98.0%
Supplier: TCI America
Synonyms:
2,5-Dimethylpyridine
L0065-25ML
L0065-500ML
L0065-5ML
TCL0065-025MLEA
187
USD
TCL0065-025ML
TCL0065-500ML
TCL0065-5ML
2,5-Lutidine ≥98.0%
2,5-Lutidine
CAS Number: 589-93-5
MDL Number: MFCD00006343
Molecular Formula: C7H9N
Molecular Weight: 107.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 157
Melting point (°C): -6
Flash Point (°C): 48
Specific Gravity (20/20): 0.93
Product Categories: Synthetic Organic Chemistry, Ligands, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: 2,5-Dimethylpyridine
Formula:
C₇H₉N MW: 107.16 g/mol Boiling Pt: 157 °C Melting Pt: -6 °C Density: 0.93 (20 °C) Flash Pt: 48 °C |
MDL Number:
MFCD00006343 CAS Number: 589-93-5 EINECS: 209-666-9 UN: 1993 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...